Order Entry
United States
ContactUsLinkComponent
Sub-X® Mounting Medium, Leica®
Sub-X® Mounting Medium, Leica®
Catalog # 89130-988
Supplier:  Avantor
CAS Number:  
undefined
Sub-X® Mounting Medium, Leica®
Catalog # 89130-988
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Sub-X® Mounting Medium
  • Size:
    473 mL (16 oz.)
  • Cat. No.:
    89130-988
  • Supplier no.:
    3801740

Specifications

About this item

This quick-drying, permanent mountant is formulated to be compatible with a Sub-X® clearing agent and all xylene substitutes on the market. It is toluene-based and may also be used to coverslip preparations cleared with xylene or toluene. Its low viscosity provides an easy flow, bubble-free medium suitable for manual or automated coverslipping. An antioxidant inhibits fading of stains.

  • Low viscosity allows for easy flowing
  • Rapid-drying
  • Toluene-based mounting medium
  • Bubble-free medium suitable for manual or automated coverslipping

It is toluene-based and may also be used to cover slip preparations cleared with xylene or toluene. Its low viscosity provides an easy flow, bubble-free medium suitable for manual or automated coverslipping. An antioxidant inhibits fading of stains.