About this item
These routine tissue cassettes are designed to keep specimens safely submerged in liquid and are resistant to the chemical action of solvents used in histology labs.
- Keep specimen submerged
- High-density polymer
- Flow-through slots
They are molded from a special high-density acetal polymer and feature efficient flow-through slots to maximize fluid exchange and ensure proper drainage. The one-piece snap-latch and hinge-lock design prevents early separation of the base and lid, and allows for one-handed operation. The lids are attached in an open position for easy filling, but can be opened or closed as often as necessary and will always relock securely. The anterior writing areas of the cassette is at 35° angles. Cassettes are specifically designed for the Leica® IP C Cassette Printer and are taped for easy loading.