RF Direct Slide Tests with Disposable Cards, Arlington Scientific
Supplier: Arlington Scientific
Certificates
About this item
The ASI RF Slide Test is a convenient and specific slide agglutination assay for the Qualitative and Semiquantitative determination of Rheumatoid Factor (RF) in human serum. No initial dilution of patient samples is required for this test.
- 2 Minute slide test
- Sensitivity 8.0 IU/ml
- Specificity >95%
- Standardized to WHO reference
- No initial dilution required
- 2 Liquid controls included
- Longest shelf life - up to 24 months
- Made in USA
- Proven by CAP, AAB, and API surveys
Certifications: CE marked. CPT Code 86430. 510(k)-K821319
Frequently Bought Together