Order Entry
United States
Orders LinkContactUsLinkComponent
VWR® Hypotonic Lysis Buffer

VWR® Hypotonic Lysis Buffer

HYPOTONIC LYSIS BUFFER, 100 mL
Catalog # 97063-130
Supplier:   VWR
Supplier #:  M334-100ML
VWR® Hypotonic Lysis Buffer
Catalog # 97063-130
Supplier:  VWR
CAS Number:  
VWR® Hypotonic Lysis Buffer
VWR® Hypotonic Lysis Buffer

HYPOTONIC LYSIS BUFFER, 100 mL

CAS Number:  
Price
$143.58
Log in for availability
Each (100ml)
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Cell lysis in detergent-free Hypotonic Lysis buffer and the subsequent isolation of the crude cytoplasmic fraction can be completed in less than 30 minutes

The protocol is compatible with a variety of downstream applications including Western Blotting and ELISA procedures as well as protein assays, which are sensitive to the presence of detergents in the lysis buffer

Documents

Specifications

Specifications

  • Description:
    HYPOTONIC LYSIS BUFFER
  • Size:
    100 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.