About this item
For more convenient gel casting, VWR® also offers Agarose I™ in 500 mg tablet form. Just add the required number of tablets to either TBE or TAE, mix until completely dissolved, heat and pour a gel. Tablets eliminate the weighing of agarose for the preparation of 0.5%, 1.0%, 1.5% and 2.0% gels.
- All-purpose, hgh purity agarose
- Exceptional band resolution and clarity
- Nuclease and protease-free
- Convenient tablet format available - no weighing required!
Agarose I™ is a standard melting/gelling agarose, suitable for routine nucleic acid analytical and preparative applications. Agarose I™ has a low EEO for shorter electrophoretic runs without compromising resolution. It is excellent for blotting techniques and general manipulations.