ContactUsLinkComponent
Lithium chloride 8 M in aqueous solution, Biotechnology Grade
Irritant
Lithium chloride 8 M in aqueous solution, Biotechnology Grade
Supplier:  VWR
Certificates
undefined
Irritant
Lithium chloride 8 M in aqueous solution, Biotechnology Grade
Supplier:  VWR
Certificates
Specification Test Results
  • DNase:
    none detected
  • Protease:
    none detected
  • RNase:
    none detected
  • Titration:
    7.7 Molar - 8.2 Molar
  • Appearance:
    Clear, colorless liquid.