Order Entry
United States
ContactUsLinkComponent
VWR® Acryl/Bis™ Acrylamide/Bis-Acrylamide Solution (40%) 37.5:1 Proteomics Grade
VWR® Acryl/Bis™ Acrylamide/Bis-Acrylamide Solution (40%) 37.5:1 Proteomics Grade
Catalog # 97064-916
Supplier:  VWR
undefined
VWR® Acryl/Bis™ Acrylamide/Bis-Acrylamide Solution (40%) 37.5:1 Proteomics Grade
Catalog # 97064-916
Supplier:  VWR
Supplier Number:  M157-500ML

Specifications

  • Description:
    ACRYL/BIS™ 37.5:1, 40% (W/V) SOLUTION
  • Size:
    500 mL
  • Environmentally Preferable:
  • Cat. No.:
    97064-916
  • Supplier no.:
    M157-500ML

Specifications

About this item

VWR® offers an extensive line of acrylamide and bis-acrylamide pre-weighed powder blends, pre-mixed stock solutions and ready-to-use solutions for customized polyacrylamide gel electrophoresis of proteins. Ultra-pure (>99.9%) acrylamide and bis-acrylamide powders provide the flexibility to prepare solutions having concentrations and ratios for all electrophoresis applications. Liquid blends minimize the handling of neurotoxic acrylamide.

  • Ultra-pure acrylamide powders ( > 99.9% )
  • High solubility, producing haze-free solutions
  • Liquid stable solutions eliminate handling hazardous powdered acrylamide
  • Reproducible & consistent results