


Acrylamide 40% in aqueous solution, Acryl 40™
Catalog # 97064-870
Supplier: VWR
CAS Number:
Specifications
- Size:500 ml
- Cat. No.:97064-870
- Supplier no.:M121-500ML
Specifications
Specification Test Results
- Acrylic Acid:0.005%
- Conductivity:5 umhos
- DNase:none detected
- Electrophoresis:Pass
- pH @25°C:6 - 7
- Protease:none detected
- RNase:none detected
Specification Test Results
About this item
:
:71.08 g/mol
UN:3426
ADR:6.1,III
:GHS06
:GHS08
:GHS07
CAS Number:79-06-1
Formula:C₃H₅NO
MDL Number:MFCD00008032