Order Entry
United States
ContactUsLinkComponent
Guanidinium hydrochloride 8 M in aqueous solution, Biotechnology Grade
Guanidinium hydrochloride 8 M in aqueous solution, Biotechnology Grade
Catalog # 97063-774
Supplier:  VWR
CAS Number:  
undefined
Guanidinium hydrochloride 8 M in aqueous solution, Biotechnology Grade
Catalog # 97063-774
Supplier:  VWR
Supplier Number:  M113-4L
CAS Number:  

Specifications

  • Size:
    4 L
  • Cat. No.:
    97063-774
  • Supplier no.:
    M113-4L

Specifications

Specification Test Results

  • Protease:
    none detected

Specification Test Results

About this item

 :
 :95.53 g/mol
CAS Number:50-01-1
Formula:NH₂C(=NH)NH₂·HCl
MDL Number:MFCD00013026