Order Entry
United States
ContactUsLinkComponent
Methanol, dry
Flammable
Toxic
Health
Methanol, dry
Catalog # 34126-312
Supplier:  GFS Chemicals
Small Business Enterprise
CAS Number:  
undefined
Flammable
Toxic
Health
Methanol, dry
Catalog # 34126-312
Supplier:  GFS Chemicals
Supplier Number:  99746
Small Business Enterprise
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Size:
    1 L
  • Cat. No.:
    34126-312
  • Supplier no.:
    161099746

Specifications

About this item

 :
 :32.04 g/mol
Merck Index:13,05984
Boiling Pt:64.6 °C (1013 hPa)
Melting Pt:–98 °C
Density:0.7918 g/cm³ (20 °C)
Flash Pt:11 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3,II
REACH:01-2119433307-44
 :GHS02
 :GHS06
 :GHS08
CAS Number:67-56-1
Formula:CH₃OH
MDL Number:MFCD00004595