Order Entry
United States
ContactUsLinkComponent
Macro-CON® Concentration System, Meridian Bioscience
Macro-CON® Concentration System, Meridian Bioscience
Catalog # 15157-028
Supplier:  Meridian
CAS Number:  
undefined
Macro-CON® Concentration System, Meridian Bioscience
Catalog # 15157-028
Supplier:  Meridian
Supplier Number:  970120
CAS Number:  

Specifications

  • Description:
    Concentration System
  • Cat. No.:
    15157-028
  • Supplier no.:
    970120

Specifications

About this item

A closed system for the concentration of eggs, larvae, and protozoa from preserved fecal specimens

The filtration unit is used in conjunction with the Para-Pak® specimen collection vial, creating a completely closed system and minimizing user exposure to potentially infectious agents. System uses the entire contents of the collection vial, reducing variation due to specimen sampling.