Order Entry
United States
ContactUsLinkComponent
CON-Trate® Concentration System, Meridian Bioscience
CON-Trate® Concentration System, Meridian Bioscience
Catalog # 15157-026
Supplier:  Meridian
CAS Number:  
undefined
CON-Trate® Concentration System, Meridian Bioscience
Catalog # 15157-026
Supplier:  Meridian
Supplier Number:  960200
CAS Number:  

Specifications

  • Description:
    Concentration System
  • Cat. No.:
    15157-026
  • Supplier no.:
    960200

Specifications

About this item

Standardized concentration system for the recovery of eggs, larvae, and protozoa from fecal material

System contains enough components for 200 tests. Components include filters, 15mL centrifuge tubes, caps, Reagent A (MucoPenX), and Reagent B (Ethyl Acetate).