



2-Mercaptoethanol ≥99%, Proteomics Grade for electrophoresis, for molecular biology
Supplier: VWR
Certificates
About this item
CAS Number:60-24-2
:
Formula:HSCH₂CH₂OH
MDL Number:MFCD00004890
Merck Index:13,05894
Boiling Pt:157.2 °C
Melting Pt:–100 deg °C
Density:1.12 g/cm³ (20 °C)
Flash Pt:68 °C
UN:2966
ADR:6.1,II
:78.14 g/mol
EINECS:200-464-6
:GHS06
:GHS08
:GHS05
:GHS07
Storage Temperature:Ambient
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- DNase:none detected
- Protease:none detected
- Purity:= 99 %
- RNase:none detected
- Water:= 0.5 %
Specification Test Results
Related Information
Frequently Bought Together