Order Entry
United States
Orders LinkContactUsLinkComponent
D-(+)-Mannose, high purity

D-(+)-Mannose, high purity

Supplier:   VWR
D-(+)-Mannose, high purity
Supplier:  VWR
Certificates
D-(+)-Mannose, high purity

1 Options Available Below

Product Details & Documents
Synonyms:  (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
  • CAS Number:3458-28-4
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00064122
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,05772
  • Boiling Pt:376 °C (1013 hPa)
  • Melting Pt:132 °C (dec.)
  • Density:1.539 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:222-392-4

Documents

Specifications for Product Family

Specification Test Results

  • Loss on Drying:
    1%
  • Melting Point:
    125°C - 131°C
  • Purity (HPLC):
    99.5 %
  • Solubility (5%, Water):
    Pass
  • Specific Rotation:
    14° - 15°
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • HDPE bottle
    100 g
    Discontinued
    Supplier Number: -
  • HDPE bottle
    500 g
    Log in for availability
    Log in for availability
    Log in for availability
    Each (500g)
    Each (500g) - $539.72
    $539.72
    Log in for availability
    Case of 4 (500g)
    Case of 4 (500g) - $2,158.82
    $2,158.82
    Each (500g)
       $539.72

    Case of 4 (500g)
       $2,158.82

    Supplier Number: J443-500G
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.