IPTG (isopropyl-β-D-thiogalactopyranoside), Ultrapure
Catalog # 97063-174
Supplier: VWR
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Bottle, amber
- Size:100 g
- Cat. No.:97063-174
- Supplier no.:J436-100G
Specifications
Specification Test Results
- Abs.@300nm (5%, Water):0.13
- Abs.@400nm (5%, Water):0.06
- Dioxane Test:Pass
- Melting Point:110°C - 114°C
- Moisture (KF):1 %
- pH (5%, Water) @25°C:5.0 - 7.0
- Purity:99 %
- Solubility (5%, Water):Pass
- Specific Rotation (1%, Water):-34.5° - -28.5°
Specification Test Results
About this item
:
:238.31 g/mol
Boiling Pt:402 °C (1013 hPa)
Melting Pt:109.5…110.5 °C
Storage Temperature:Freezer
EINECS:206-703-0
CAS Number:367-93-1
Formula:C₉H₁₈O₅S
MDL Number:MFCD00063273