Order Entry
United States
Orders LinkContactUsLinkComponent
 

HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid free acid, high purity for biochemistry

Supplier:   VWR
HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid free acid, high purity for biochemistry
Supplier:  VWR
Certificates
HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid free acid, high purity for biochemistry
HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid free acid, high purity for biochemistry

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

One of the best general-purpose buffers for biological research. Excellent for tissue culture, oxidative phosphorylation, protein synthesis with cell-free bacterial systems, photophosphorylation, and CO2 fixation.

Suitable buffer for TEM studies (Transmission Electron Microscopy) as it does not affect metal substrates. Recommended buffer for the glutamate binding assay, prevents binding to non-receptor materials.

Synonyms:  HEPES free acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulfonic acid, 4-(2-Hydroxyethyl)piperazin-1-ylethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid), 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid
  • CAS Number:7365-45-9
  • Formula:C₈H₁₈N₂O₄S
  • MDL Number:MFCD00006158
  • Molecular Weight:238.31 g/mol
  • Merck Index:13,04672
  • Melting Pt:234 °C
  • Density:1.448 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:230-907-9
Specifications for Product Family

Specification Test Results

  • DNase:
    none detected
  • Heavy Metals (as Pb):
    = 0.0005 %
  • Iron:
    = 0.0005 %
  • pKa @20°C:
    7.45 - 7.65
  • Protease:
    none detected
  • Purity (Dry Basis):
    99 %
  • Residue on Ignition:
    0.2 %
  • RNase:
    none detected
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • HDPE bottle
    50 g
    Log in for availability
    Log in for availability
    Log in for availability
    Each (50g)
    Each (50g) - $93.37
    $93.37
    Log in for availability
    Case of 12 (50g)
    Case of 12 (50g) - $841.60
    $841.60
    Each (50g)
       $93.37

    Case of 12 (50g)
       $841.60

    Supplier Number: 0511-50G
  • HDPE bottle
    250 g
    Log in for availability
    Log in for availability
    Log in for availability
    Each (250g)
    Each (250g) - $271.01
    $271.01
    Log in for availability
    Case of 12 (250g)
    Case of 12 (250g) - $2,640.71
    $2,640.71
    Each (250g)
       $271.01

    Case of 12 (250g)
       $2,640.71

    Supplier Number: 0511-250G
  • HDPE bottle
    1 kg
    Log in for availability
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1kg)
    Each (1kg) - $722.33
    $722.33
    Log in for availability
    Case of 4 (1kg)
    Case of 4 (1kg) - $2,505.52
    $2,505.52
    Log in for availability
    Pallet of 240 (1kg)
    Pallet of 240 (1kg) - $150,330.65
    $150,330.65
    Each (1kg)
       $722.33

    Case of 4 (1kg)
       $2,505.52

    Pallet of 240 (1kg)
       $150,330.65

    Supplier Number: 0511-1KG

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.