Order Entry
United States
Orders LinkContactUsLinkComponent
N,N-Dimethylformamide, Reagent Grade

N,N-Dimethylformamide, Reagent Grade

Bottle, amber, 500 ml
Catalog # 97064-586
Flammable
Health
Irritant
Supplier:   VWR
Supplier #:  0464-500ML
N,N-Dimethylformamide, Reagent Grade
Catalog # 97064-586
Supplier:  VWR
CAS Number:  68-12-2
N,N-Dimethylformamide, Reagent Grade
N,N-Dimethylformamide, Reagent Grade

Bottle, amber, 500 ml

CAS Number:  68-12-2
Price
$56.91
Log in for availability
Each (500ml) Case of 6 (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Formic acid dimethylamide, DMFA, Formyldimethylamine, N-Formyldimethylamine, Dimethylformamide, NSC 5356, DMF
  • CAS Number:68-12-2
  • Formula:HCON(CH₃)₂
  • MDL Number:MFCD00003284
  • Molecular Weight:73.09 g/mol
  • Merck Index:13,03269
  • Boiling Pt:153 °C (1013 hPa)
  • Melting Pt:–61 °C
  • Density:0.949 g/cm³ (20 °C)
  • Flash Pt:57.5 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-679-5
  • UN:2265
  • ADR:3,III
  • REACH:01-2119475605-32

Documents

Specifications

Specification Test Results

  • Refractive Index:
    1.423 - 1.431

Specifications

  • Pack type:
    Bottle, amber
  • Size:
    500 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.