Order Entry
United States
ContactUsLinkComponent
KIMBERLY-CLARK PROFESSIONAL* WYPALL* Jumbo Roll Dispenser
KIMBERLY-CLARK PROFESSIONAL* WYPALL* Jumbo Roll Dispenser
Catalog # 500043-385
undefined
KIMBERLY-CLARK PROFESSIONAL* WYPALL* Jumbo Roll Dispenser
Catalog # 500043-385
Supplier Number:  KCC80596

Specifications

  • Color:
    Black
  • Cat. No.:
    500043-385
  • W×D×H:
    16 3/4 × 18 1/2 × 33''
  • For use with:
    Jumbo Rag-On-A-Roll* System
  • Supplier no.:
    KCC80596

Specifications

About this item

A mobile, efficient dispensing system that reduces waste

Dispenser for JUMBO RAG-ON-A-ROLL* system. For Jumbo Rolls KIM-41055, KIM-41600 and KIM-34955 (sold separately). Portable, high-capacity dispenser is compact enough for use in most any location. Metal with trash bag attachment.