Order Entry
United States
ContactUsLinkComponent
KIMBERLY-CLARK PROFESSIONAL* IN-SIGHT* Roll Control Center Pull Dispenser
KIMBERLY-CLARK PROFESSIONAL* IN-SIGHT* Roll Control Center Pull Dispenser
Catalog # 500008-500
undefined
KIMBERLY-CLARK PROFESSIONAL* IN-SIGHT* Roll Control Center Pull Dispenser
Catalog # 500008-500
Supplier Number:  KCC09989

Specifications

  • Color:
    Translucent Smoke/Gray
  • Cat. No.:
    500008-500
  • W×D×H:
    10.2 × 9 1/2 × 11.8"
  • For use with:
    One 8" and One 3.5" Diameter Roll
  • Supplier no.:
    KIM09989

Specifications

About this item

Higher capacity, reduced maintenance system

Easy to maintain—one-handed loading, plus a patented aperture that reduces dispensing problems. Use with SCOTT® Towels KIM-01032 (sold separately). Locking plastic case.