Universal® Extra Strength Easy Assembly Storage Boxes
Supplier: Janitorial Supplies
Universal® Extra Strength Easy Assembly Storage Boxes
Supplier: Janitorial Supplies
Certificates
About this item
Double wall construction on all panels increase stacking strength and portability.
Designed to stack two on a shelf. Smooth rolled edges eliminate paper cuts. Large tote handles for portability.
2 Options Available Below
Product Details & Documents
Double wall construction on all panels increase stacking strength and portability.
Designed to stack two on a shelf. Smooth rolled edges eliminate paper cuts. Large tote handles for portability.
Specifications for Product Family
Specifications
- Catalog No:
- 500003-158
- 500003-160
- Environmentally Preferable:
- Sustainable Materials
- Sustainable Materials
Specifications
Product Family Options
Product Information
- ColorMaterialStacking strengthSizeDimensionsAvailabilityPrice
- 500003-158with Lift-Off Lidwith Lift-Off LidWhiteCorrugated Fiberboard700 lbs.Legal15×24×10" (Interior)Log in for availabilityCarton of 12$141.04Supplier Number: UNV95221Specifications:
Sustainable Materials More
Specifications
Cat. No.500003-158Environmentally PreferableSustainable MaterialsSustainable Materials - 500003-160with Lift-Off Lidwith Lift-Off LidWhiteCorrugated Fiberboard710 lbs.Letter12×24×10" (Interior)Log in for availabilityCarton of 12$108.85Supplier Number: UNV95220Specifications:
Sustainable Materials More
Specifications
Cat. No.500003-160Environmentally PreferableSustainable MaterialsSustainable Materials