deflect-o® Horizontal Business Card Holder, Essendant
Catalog # 500040-699
Supplier: Janitorial Supplies
Specifications
- For Use With:Business cards
- Capacity:50
- Color:Clear
- Description:Business card holder
- Dimensions:3.75×1.5×1.875"
- Material:Plastic
- Cat. No.:500040-699
- Supplier no.:DEF70101
Specifications
About this item
Tasteful, contemporary design
Durable plastic construction.