Sharpie® T.E.C. Permanent Marker
Black
Catalog # 500039-912
Supplier: Janitorial Supplies
Supplier #: SAN13401
Jump to:
Product Details & Documents
Fine-tip permanent marker marks on most surfaces.
Meets requrements for trace element certified ink for the nuclear energy, aerospace, electronic, aeronautic and ship building industries. Each marker is identified with the batch number of the ink.
Specifications
- Color:Black