3M™ Red Buffer Floor Pads 5100
Catalog # 500039-344
Supplier: Janitorial Supplies
Specifications
- Ø:19"
- Color:Red
- Material:Non-Woven Polyester Fiber
- Cat. No.:500039-344
- Supplier no.:MMM08394
Specifications
About this item
Specially designed for spray buffing.
Cleans when damp; buffs when dry. Quickly cleans and removes scuff marks and enhances floor appearance. Works great on automatic floor scrubbers for light-duty cleaning.
Used with rotary or automatic machine speed of 175 to 600 rpm.