Dataproducts® R0910 Cash Register Ribbon, Essendant
Catalog # 500038-526
Supplier: Janitorial Supplies
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Color:Purple
- Description:Cash register ribbon for IR91P, nylon, compatible, 2 7/8"
- Cat. No.:500038-526
- Supplier no.:DPSR0910
Specifications
About this item
For use with Citizen® MD 910 and 911 cash registers
Consistent, crisp character output.