8200 N95 Particle Respirator 8200, 3M™
Catalog # 500023-588
Supplier: Janitorial Supplies
Specifications
- Description:8200 N95 Particle Respirator
- Cat. No.:500023-588
- Supplier no.:MMM8200
Specifications
About this item
These lightweight respirators protect the workers and co-workers from work around tiny particles.
- Resistant to airborne germs, dust, bodily fluids
- Adjustable nose clip ensures a more secure seal against the face
- Advanced electrostatically charged microfibers help make breathing easier
At least 95% filtration efficiency when subjected to a 0.3 µm MMAD sodium chloride particle challenge. Suggested for applications such as bagging, sanding and sweeping.
Caution: Must be used in accordance with OSHA regulations, and the user instructions,, warnings, and limitations that accompany each item.