Universal® 100% Recycled Hardboard Clipboard with High-Capacity Clip
Catalog # 500006-290
Supplier: Janitorial Supplies
Specifications
- Color:Brown
- Description:Hardboard, clipboard, holds paper size 5x8", clip capacity 1"
- Cat. No.:500006-290
- W×H:6×9"
- Supplier no.:UNV05610
Specifications
About this item
Made from the finest hardboard available—smooth front and back surfaces eliminate unsightly hardboard "dust." High-capacity clip.