Order Entry
United States
ContactUsLinkComponent
Innovera® Wireless Optical Mouse with Micro USB, Essendant LLC MS
Innovera® Wireless Optical Mouse with Micro USB, Essendant LLC MS
Catalog # 500017-394
undefined
Innovera® Wireless Optical Mouse with Micro USB, Essendant LLC MS
Catalog # 500017-394
Supplier Number:  IVR61025

Specifications

  • Color:
    Gray/Black
  • Cat. No.:
    500017-394
  • Supplier no.:
    IVR61025

Specifications

About this item

Improved optical function! New optical technology allows mouse to track on most surfaces.

  • 2 AAA battery
  • USB connector and Wireless
  • Left/Right hand use
  • For use with Windows® 2000, XP, Vista™, 7

No more productivity slow-downs while you wait to find a tracking surface. An ideal solution for desktop use and traveling, this wireless mouse has a micro USB receiver that can be left plugged into a laptop. DPI button allows switching between 800/1600 dpi for more precise control when you need it. Rubber grips and an ergonomic shape make the mouse comfortable to use.