Order Entry
United States
ContactUsLinkComponent
 
Tabbies® Medical Chart Index Divider Sheets
  500044-553
undefined
Tabbies® Medical Chart Index Divider Sheets
  500044-553
 :  TAB54520

 

  • For Use With:
    Three-Ring Binder, Two-Prong Fastener Folder
  • Punched/unpunched:
    7-hole punched
  • Color:
    White
  • Description:
    Untabbed divider
  • Material:
    Paper, 110-lb.
  • Size:
    11 x 8 1/2"
  • Cat. No.:
    500044-553
  • Supplier no.:
    TAB54520
  • Tab style:
    Untabbed

 

 

Divider sheets are 110-lb

stock printed with guidelines for fast and accurate placement of medical chart divider index tabs. Sheets are conveniently ruled for making notes and are 7-hole punched to be used with fasteners in folders or 3-ring binders. Orientation: portrait.