Order Entry
United States
ContactUsLinkComponent
 
Logitech® M510 Wireless Mouse, Essendant LLC MS
  500041-016
 :  Avantor
undefined
Logitech® M510 Wireless Mouse, Essendant LLC MS
  500041-016
 :  Avantor

 

  • Color:
    Dark Gray
  • Height:
    2¼"
  • Cat. No.:
    500041-016
  • Depth:
    9"
  • Width:
    4⅗"
  • Supplier no.:
    LOG910001822

 

 

Get the ergonomic support you need with this wireless mouse that features soft rubber grips, gently curved sides and broad palm area.

  • Advanced 2.4 GHz wireless connection
  • Right hand use
  • For use with Mac® OS X 10.4 and Later, Windows® XP, Vista™, 7

Standard and programmable buttons make flipping through photo albums and documents a breeze. Logitech® Unifying USB receiver stays plugged into your laptop for added versatility and is compatible with other Unifying mice and keyboards. Side-to-side scrolling allows you to quickly navigate through your music library, favorite web pages and more. On/off switch and smart sleep mode help conserve and extend battery life.