Order Entry
United States
ContactUsLinkComponent
Sealed Air Bubble Wrap® Cushioning Material
Sealed Air Bubble Wrap® Cushioning Material
Catalog # 500030-732
undefined
Sealed Air Bubble Wrap® Cushioning Material
Catalog # 500030-732
Supplier Number:  SEL90065

Specifications

  • Description:
    Packaging with bubble cushioning type
  • Length:
    65'
  • Thickness:
    1.3 cm (0.5")
  • Cat. No.:
    500030-732
  • Width:
    12"
  • Supplier no.:
    SEL90065

Specifications

About this item

Bubble Wrap® provides the quality and performance for all shipping and packing needs.

This packaging helps preserve products by creating an inbarrier layer for protection during transit or storage. Made with a co-extruded barrier that is 100 times more resistant to the passage of air than even a thick single layer of polyethylene.