Velcro® One-Wrap® Reusable Ties
Catalog # 500009-858
Supplier: Janitorial Supplies
Specifications
- Color:Black, Gray
- Description:Fastener
- Dimensions:1/2x8" per pad
- Application:For bundling cords
- Cat. No.:500009-858
- Supplier no.:VEK90924
Specifications
About this item
Take control of cord clutter
Bundle computer and electronics cords with these easily adjustable, reusable ties. The simple one-piece self-gripping design wraps onto itself for a secure hold. Lightweight, low profile tie attaches to cords to prevent loss when objects are unbundled.