LYSOL® Brand I.C.™ Foaming Disinfectant Cleaner
Catalog # 500032-618
Supplier: Avantor
Specifications
- Description:Foam Aerosol Disinfectant, fresh scented
- Size:24 oz.
- Cat. No.:500032-618
- Supplier no.:RAC95524EA
Specifications
About this item
Ready-to-use quaternary formula cleans, disinfects and deodorizes hard, nonporous surfaces such as tile, fiberglass, marble, plastic, porcelain and vinyl
- Diethylene Glycol Monobutyl Ether compound
- Contains no harsh acids, abrasives or bleach
Kills Pseudomonas, Salmonella, Staph, E.coli, Tuberculosis (TB), HIV-1 (AIDS), Hepatitis A, Herpes simplex 1 and 2, Influenza A2, Polio and RSV viruses, among others.
Applicable on fiberglass, marble, porcelain and vinyl.
Certifications: EPA Registered, NSF Category Code C2. Meets AOAC efficacy standards for hospital aerosol disinfectants.