Order Entry
United States
ContactUsLinkComponent
Elmer's® Multi-Purpose Spray Adhesive
Elmer's® Multi-Purpose Spray Adhesive
Catalog # 500031-592
undefined
Elmer's® Multi-Purpose Spray Adhesive
Catalog # 500031-592
Supplier Number:  EPIE451

Specifications

  • Volume:
    11 oz.
  • Color:
    Clear
  • Description:
    Adhesive spray, aerosol
  • Environmentally Preferable:
  • Application:
    Applicable to a variety of surfaces including plastics
  • Cat. No.:
    500031-592
  • Packaging:
    Can
  • Supplier no.:
    EPIE451

Specifications

About this item

Specially formulated for mounting items to a variety of surfaces, including plastics.

  • Dries fast and clear
  • Sprays wide
  • Acid-free

Delivers fast-grab mounting to styrofoam and foam board. Dries fast and clear, resisting bleedthrough, to create a permanent bond. Sprays wide to cover large areas.