Order Entry
United States
ContactUsLinkComponent
Universal® Retractable Pen-Style Correction Tape
Universal® Retractable Pen-Style Correction Tape
Catalog # 500026-920
Supplier:  Avantor
undefined
Universal® Retractable Pen-Style Correction Tape
Catalog # 500026-920
Supplier:  Avantor

Specifications

  • Color:
    White
  • Description:
    Correction tape
  • Environmentally Preferable:
  • Cat. No.:
    500026-920
  • Packaging:
    4/pack
  • Supplier no.:
    UNV75605
  • Tape length:
    236"
  • Tape width:
    0.20"

Specifications

About this item

Pen-style correction tape dispenser is ergonomically designed to fit your hand for maximum comfort

Two-way function provides flexibility—pull the applicator to the right for correcting whole sentences, or push to the left if you only need to correct a single letter. Retractable tip cover protects tape when not in use. Great for left- or right-handed use.