Order Entry
United States
ContactUsLinkComponent
 
HON® Center Drawer for Double Pedestal Desks, Essendant LLC MS
Catalog # 500008-740
   Sustainable Materials
   Safer Human and Environmental Health
undefined
HON® Center Drawer for Double Pedestal Desks, Essendant LLC MS
Catalog # 500008-740
Supplier Number:  HOND8S
   Sustainable Materials
   Safer Human and Environmental Health

Specifications

  • Depth:
    14¾"
  • Width:
    24¾"
  • Color:
    Charcoal
  • Height:
    3"
  • Material:
    Metal
  • Environmentally Preferable:
    Sustainable Materials,
    Safer Human and Environmental Health
  • Environmental Certification:
    level® 2 Certified, Scientific Certification Systems (SCS) Certified Indoor Advantage™ Gold
  • Cat. No.:
    500008-740
  • For use with:
    Double pedestal desk HON38180ML
  • Supplier no.:
    HOND8S

Specifications

About this item

Organize your supplies by adding this center drawer to a Double Pedestal Desk.

  • Radius edge

Core removable lock.

Certifications: Meets or exceeds ANSI/BIFMA standards.