Canon® Ink Cartridge, 2950B001, Essendant LLC MS
Catalog # 500014-416
Supplier: Janitorial Supplies
Specifications
- Cartridge no.:CLI-221
- For Use With:Inkjet Printer
- Color:Gray
- Cat. No.:500014-416
- Supplier no.:CNM2950B001
Specifications
About this item
An ink cartridge that redefines the benchmark for color, durability and longevity.
- OEM and not remanufactured
Receive exceptional color while increasing the profitability per print. Choose this cartridge when the printing requirements are most demanding.