Order Entry
United States
ContactUsLinkComponent
Alera® Wire Shelving Shelf Lock Clips, Essendant LLC MS
Alera® Wire Shelving Shelf Lock Clips, Essendant LLC MS
Catalog # 500016-819
undefined
Alera® Wire Shelving Shelf Lock Clips, Essendant LLC MS
Catalog # 500016-819
Supplier Number:  ALESW59SLBL

Specifications

  • For Use With:
    Alera® Wire Shelving Units
  • Color:
    Black
  • Material:
    Plastic
  • Cat. No.:
    500016-819
  • Supplier no.:
    ALESW59SLBL

Specifications

About this item

Locks down shelving securely

Constructed to stand up to daily wear-and-tear.

Certifications: GREENGUARD Children & Schools Certified, Meets or exceeds ANSI/BIFMA standards.