Order Entry
United States
ContactUsLinkComponent
deflect-o® Stackable Wall Pocket, Essendant
deflect-o® Stackable Wall Pocket, Essendant
Catalog # 500038-481
undefined
deflect-o® Stackable Wall Pocket, Essendant
Catalog # 500038-481
Supplier Number:  DEF74301

Specifications

  • Color:
    Clear
  • Description:
    Wall-mountable file pocket, file size format: legal
  • Height:
    7"
  • Material:
    Plastic
  • Environmentally Preferable:
  • Cat. No.:
    500038-481
  • Depth:
    4"
  • Width:
    16¼"
  • Supplier no.:
    DEF74301

Specifications

About this item

Keep files within easy reach with stackable pockets

Mounts easily to wall with screws or included mounting tape. Additional pockets (not included) stack onto each other so you can expand your storage without adding more holes to your walls.