Order Entry
United States
ContactUsLinkComponent
Stanley Bostitch® Xtreme Duty Adjustable Paper Punch with Antimicrobial Protection, Essendant LLC MS
Stanley Bostitch® Xtreme Duty Adjustable Paper Punch with Antimicrobial Protection, Essendant LLC MS
Catalog # 500022-790
undefined
Stanley Bostitch® Xtreme Duty Adjustable Paper Punch with Antimicrobial Protection, Essendant LLC MS
Catalog # 500022-790
Supplier Number:  BOS03200

Specifications

  • Sheet capacity:
    160
  • Color:
    Black/Silver
  • Description:
    Metal, extra heavy-duty hole punch
  • Cat. No.:
    500022-790
  • Hole diameter:
    ⁹⁄₃₂"
  • No. of holes:
    2 - 3
  • Supplier no.:
    BOS03200

Specifications

About this item

Antimicrobial compound safely reduces the growth of bacteria and mold.

  • Punches two or three holes
  • Manual operating
  • Round shaped punch

Precision hollow steel punch heads for clean punching of up to 160 sheets of 20 lb. paper with minimal effort. Easy clean clip tray maintains a clean work environment.

Caution: Sheet capacities are based on 20 lbs. bond. As more heads are added to a punch, the sheet punching capacity decreases.