Waxed Kraft Liners, Hospital Specialty Co.
500016-638
- Color:Kraft
- Cat. No.:500016-638
- W×D×H:10 × 3 1/4 × 9"
- Supplier no.:HOS6141
For use with wall and floor-type sanitary napkin receptacles (sold separately).
For use with wall and floor-type sanitary napkin receptacles (sold separately).