Order Entry
United States
ContactUsLinkComponent
Innovera® Drum Unit, DR350, Essendant LLC MS
Innovera® Drum Unit, DR350, Essendant LLC MS
Catalog # 500027-845
undefined
Innovera® Drum Unit, DR350, Essendant LLC MS
Catalog # 500027-845
Supplier Number:  IVRDR350
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • For Use With:
    Fax
  • Yield:
    12000 pages
  • Color:
    Black
  • Environmentally Preferable:
  • Cat. No.:
    500027-845
  • Supplier no.:
    IVRDR350

Specifications

About this item

Rely on genuine OEM quality without the OEM price.

  • Compatible
  • Remanufacturing available

Produces clear, sharp and professional printing results. Simple installation means greater efficiency.