Alera® Core Removable Lock and Key Set, Essendant LLC MS
Catalog # 500016-809
Supplier: Janitorial Supplies
Specifications
- For Use With:Alera® Valencia Series
- Color:Silver
- Description:Key Lockout
- Material:Steel
- Cat. No.:500016-809
- Supplier no.:ALEVA501111
Specifications
About this item
Keep valuable contents safe and secure
Replaces existing locks with keyed-alike locks in all storage units. Each order received for multiple locks will be shipped keyed-alike. Order one core/key set for each storage unit.
Certifications: GREENGUARD Indoor Air Quality Certified®, Meets or exceeds ANSI/BIFMA standards.
Ordering information: Number of keys 2.