Acroprint® ETC Time Stamp Clock Ribbon, Essendant
Catalog # 500024-064
Supplier: Janitorial Supplies
Specifications
- Color:Purple
- Description:OEM ribbon for time clock
- Material:Polyester/steel
- Cat. No.:500024-064
- Supplier no.:ACP200137000
Specifications
About this item
For use with Acroprint® Models ET and ETC time stamps
Accurate, clear output.