Acroprint® ES700 and ES900 Electronic Payroll Recorder/Time Stamp/Numbering Machine, Backup Time Clock Ribbon, Essendant
Catalog # 500024-051
Supplier: Janitorial Supplies
Specifications
- Color:Black
- Description:OEM ribbon for time clock ES 700/900
- Material:Polyester/plastic
- Cat. No.:500024-051
- Supplier no.:ACP390121000
Specifications
About this item
For use with Acroprint® payroll recorder model ES900
Accurate, clear output.