Brita® Pitcher Replacement Filters
Catalog # 500010-957
Supplier: Janitorial Supplies
Specifications
- Description:Pitcher Filter
- Cat. No.:500010-957
- Supplier no.:CLO35503
Specifications
About this item
Ensure your drinking water is fresh, great tasting and healthier! Reduces chlorine (taste and odor), lead, copper and mercury content (substances reduced may not be in all users' water)
Replace every 40 gallons (approximately two months). Each filter saves up to 300 bottles of water.