ContactUsLinkComponent
Brita® Faucet Filter System
Brita® Faucet Filter System
  500017-382
undefined
Brita® Faucet Filter System
  500017-382
 :  CLO42201
  • Cat. No.:
    500017-382
  • Supplier no.:
    CLO42201
Brita's multistage faucet filter reduces impurities and has an added layer to trap sediment
All that's left behind is cleaner, great-tasting water. Reduces Lead, Cryptosporidium and Giardia, TTHMs and VOC (substances reduced may not be in all users´ water). Plus this space-saving filtration system has advanced features for the ultimate in convenience. Its long-lasting filter and easy access make this the perfect option for families, kitchens and cooking. Convenient spray option gives you control over 3 flow options: filtered water, spray or stream. Electronic filter-change indicator tells you when to change your filter. Easy installation—attaches to faucet in minutes without tools. White.