Order Entry
United States
ContactUsLinkComponent
Dataproducts® R4000 Printer Ribbon, Essendant
Dataproducts® R4000 Printer Ribbon, Essendant
Catalog # 500023-912
undefined
Dataproducts® R4000 Printer Ribbon, Essendant
Catalog # 500023-912
Supplier Number:  DPSR4000

Specifications

  • For Use With:
    Epson® DFX5000, 5000+, 8000 , 8500 printers
  • Color:
    Black
  • Description:
    Compatible nylon printer ribbon
  • Environmentally Preferable:
  • Cat. No.:
    500023-912
  • Supplier no.:
    DPSR4000

Specifications

About this item

This printer Ribbon has long life and exceptional character definition.

  • Used for dot matrix printer