Belkin® CAT5e Patch Cables
About this item
Premium patch cable delivers reliable performance in 10/100Base-T networking applications
Fifty-micron, gold-plated connectors ensure clean, clear transmissions. Rigorous PowerSUM testing is designed to exceed Cat5e standard performance requirements.
1 Options Available Below
Product Details & Documents
Premium patch cable delivers reliable performance in 10/100Base-T networking applications
Fifty-micron, gold-plated connectors ensure clean, clear transmissions. Rigorous PowerSUM testing is designed to exceed Cat5e standard performance requirements.
Product Family Options
Product Information
- ColorLengthAvailabilityPrice
- 500003-646Cat5e Patch molded cable for Mac®, network, PC 2x RJ 45 maleCat5e Patch molded cable for Mac®, network, PC 2x RJ 45 maleGrey7.6 m (25 ft.)Log in for availabilityEach$15.72Supplier Number: BLKA3L79125M
- Discontinued-Supplier Number: -Specifications:MarketsourceMarketsource