Order Entry
United States
ContactUsLinkComponent
Rubbermaid® Commercial Brute® Recycling Container
Rubbermaid® Commercial Brute® Recycling Container
Catalog # 500023-062
undefined
Rubbermaid® Commercial Brute® Recycling Container
Catalog # 500023-062
Supplier Number:  RCP263273BE

Specifications

  • Ø:
    22"
  • Width:
    22"
  • Capacity:
    32 gal.
  • Color:
    Blue
  • Description:
    Open top recycling receptacle
  • Height:
    27¼"
  • Material:
    Plastic
  • Cat. No.:
    500023-062
  • Shape:
    Round
  • For use with:
    Lid RCP-263100BE (sold separately)
  • Supplier no.:
    RCP263273BE

Specifications

About this item

You'll be happy to have this Brute® on your side.

Sturdy 32-gallon receptacle can handle double duty as central collection point and shipping container. Reduces the amount of waste staff handles waste. Transport waste with ease and efficiency.