Order Entry
United States
ContactUsLinkComponent
Scotch® Box Sealing Tape Dispenser
Scotch® Box Sealing Tape Dispenser
Catalog # 500043-712
undefined
Scotch® Box Sealing Tape Dispenser
Catalog # 500043-712
Supplier Number:  MMM37502ST

Specifications

  • For Use With:
    Tape width: 2"; tape length: 60 yds.
  • Color:
    Gray
  • Description:
    Tape with dispenser, value pack, core size 3", with handle
  • Cat. No.:
    500043-712
  • Supplier no.:
    MMM37502ST

Specifications

About this item

This lightweight, roll-on tape dispenser with pistol grip handle allows you to attach, apply and cut tape with only one hand.

Features an adjustable tension that controls tape unwind--helping you to save time and money. The built-in adjustable break provides a tight, consistent seal. Made of metal and gray high-impact plastic for durability and lasting performance. Easy to load design.

Certifications: Meets U.S. Postal Regulations for standard packages.